siRNA Details
Gene Symbol: Rab23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109005.1 | NP_001102475.1 | 6 | 796 | ||
XM_008766955.1 | XP_008765177.1 | 6 | 1457 | |||
XM_008766956.1 | XP_008765178.1 | 6 | 1462 | |||
XM_008766957.1 | XP_008765179.1 | 6 | 782 | |||
XM_008766958.1 | XP_008765180.1 | 6 | 1124 | |||
XM_008766959.1 | XP_008765181.1 | 6 | 1455 | |||
XM_008766960.1 | XP_008765182.1 | 6 | 1450 | |||
XM_008766961.1 | XP_008765183.1 | 4 | 2173 |
