siRNA Details
Gene Symbol: Rab23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109005.1 | NP_001102475.1 | 8 | 985 | ||
XM_008766955.1 | XP_008765177.1 | 8 | 1646 | |||
XM_008766956.1 | XP_008765178.1 | 8 | 1651 | |||
XM_008766957.1 | XP_008765179.1 | 8 | 971 | |||
XM_008766958.1 | XP_008765180.1 | 8 | 1313 | |||
XM_008766959.1 | XP_008765181.1 | 8 | 1644 | |||
XM_008766960.1 | XP_008765182.1 | 8 | 1639 | |||
XM_008766961.1 | XP_008765183.1 | 6 | 2362 |
