siRNA Details
Gene Symbol: LOC367746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773102.1 | XP_008771324.1 | 2 | 726 | ||
GenBank mRNA | BC086373.1 | - | 2 | 633 |
Gene Symbol: LOC367333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767409.1 | XP_008765631.1 | 2 | 656 |
