siRNA Details
Gene Symbol: Tceanc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109015.1 | NP_001102485.1 | 1 | 365 | ||
XM_006256856.2 | XP_006256918.1 | 5 | 1239 | |||
XM_008773182.1 | XP_008771404.1 | 5 | 676 | |||
XM_008773183.1 | XP_008771405.1 | 5 | 929 |
