siRNA Details
Gene Symbol: Pnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109023.1 | NP_001102493.1 | 4, 5 | 355 | ||
GenBank mRNA | FQ234501.1 | - | 4, 5 | 364 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109023.1 | NP_001102493.1 | 4, 5 | 355 | ||
GenBank mRNA | FQ234501.1 | - | 4, 5 | 364 |