siRNA Details
Gene Symbol: Pcdha7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199507.1 | NP_955801.1 | 1 | 1603 | ||
GenBank mRNA | AB113390.1 | - | 1 | 1603 | ||
AY573981.1 | - | 1 | 1603 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pcdha7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199507.1 | NP_955801.1 | 1 | 1603 | ||
GenBank mRNA | AB113390.1 | - | 1 | 1603 | ||
AY573981.1 | - | 1 | 1603 |