siRNA Details
Gene Symbol: Hyal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100780.1 | NP_001094250.1 | 3 | 256 | ||
XM_008762799.1 | XP_008761021.1 | 2 | 284 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hyal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100780.1 | NP_001094250.1 | 3 | 256 | ||
XM_008762799.1 | XP_008761021.1 | 2 | 284 |