siRNA Details
Gene Symbol: Olr695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000628.1 | NP_001000628.1 | 1 | 114 |
Gene Symbol: LOC103690375 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762021.1 | XP_008760243.1 | 114 |
