siRNA Details
Gene Symbol: Olr1523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000713.1 | NP_001000713.1 | 819 | |||
GenBank mRNA | X89694.1 | - | 445 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000713.1 | NP_001000713.1 | 819 | |||
GenBank mRNA | X89694.1 | - | 445 |