siRNA Details
Gene Symbol: Olr1288 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000798.1 | NP_001000798.1 | 1 | 630 |
Gene Symbol: LOC100911127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770725.1 | XP_008768947.1 | 1 | 630 |
