siRNA Details
Gene Symbol: Olr1292 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000960.1 | NP_001000960.1 | 1 | 675 |
Gene Symbol: LOC100910719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770724.1 | XP_008768946.1 | 1 | 675 |
