siRNA Details
Gene Symbol: Clic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002807.2 | NP_001002807.1 | 6 | 844 | ||
XM_008772745.1 | XP_008770967.1 | 6 | 849 | |||
GenBank mRNA | BC099823.1 | - | 6 | 796 | 7932557 | |
EU148064.1 | - | 6 | 608 | |||
FM115459.1 | - | 4 | 466 | |||
FQ219968.1 | - | 6 | 834 | |||
FQ227136.1 | - | 6 | 630 | |||
FQ227567.1 | - | 6 | 621 | |||
FQ227951.1 | - | 6 | 620 | |||
FQ233892.1 | - | 6 | 621 |
