siRNA Details
Gene Symbol: Pan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008862.1 | NP_001008862.1 | 16, 17 | 2480 | ||
XM_006240783.2 | XP_006240845.1 | 17, 18 | 3663 | |||
XM_006240784.2 | XP_006240846.1 | 17, 18 | 3660 | |||
XM_006240785.2 | XP_006240847.1 | 17, 18 | 3648 | |||
XM_006240786.2 | XP_006240848.1 | 17, 18 | 3651 | |||
XM_006240787.2 | XP_006240849.1 | 17, 18 | 3648 |
