siRNA Details
Gene Symbol: Pan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008862.1 | NP_001008862.1 | 4 | 593 | ||
XM_006240783.2 | XP_006240845.1 | 5 | 1761 | |||
XM_006240784.2 | XP_006240846.1 | 5 | 1761 | |||
XM_006240785.2 | XP_006240847.1 | 5 | 1761 | |||
XM_006240786.2 | XP_006240848.1 | 5 | 1761 | |||
XM_006240787.2 | XP_006240849.1 | 5 | 1761 | |||
XM_006240788.2 | XP_006240850.1 | 5 | 1761 | |||
XM_006240789.2 | XP_006240851.1 | 5 | 1761 |
