siRNA Details
Gene Symbol: Prss57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003956.1 | NP_001003956.1 | 3 | 327 | ||
XM_006240979.2 | XP_006241041.1 | 3 | 470 | |||
XM_008765175.1 | XP_008763397.1 | 3 | 470 |
