siRNA Details
Gene Symbol: Tesb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113183.1 | NP_001106654.1 | 18, 19 | 629 | ||
XM_008772754.1 | XP_008770976.1 | 27, 28 | 1217 | |||
XM_008772755.1 | XP_008770977.1 | 27, 28 | 1319 | |||
XM_008772756.1 | XP_008770978.1 | 26, 27 | 1094 | |||
XM_008772757.1 | XP_008770979.1 | 26, 27 | 1196 | |||
XM_008772758.1 | XP_008770980.1 | 26, 27 | 1196 | |||
XM_008772759.1 | XP_008770981.1 | 24, 25 | 1154 | |||
XM_008772760.1 | XP_008770982.1 | 21, 22 | 1043 | |||
GenBank mRNA | BC083628.1 | - | 21, 22 | 1205 | ||
BC097390.1 | - | 18, 19 | 629 | 7453125 |
