siRNA Details
Gene Symbol: Atp6v0e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002253.1 | NP_001002253.1 | 2, 3 | 229 | ||
GenBank mRNA | BC089958.1 | - | 181 | 7312897 | ||
CB566985.1 | - | 2, 3 | 252 | |||
CB757317.1 | - | 2, 3 | 213 | |||
FQ212114.1 | - | 1319 |
