siRNA Details
Gene Symbol: Ly96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024279.1 | NP_001019450.1 | 474 | |||
GenBank mRNA | AY963291.1 | - | 474 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ly96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024279.1 | NP_001019450.1 | 474 | |||
GenBank mRNA | AY963291.1 | - | 474 |