siRNA Details
Gene Symbol: Omg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005898.3 | NP_001005898.3 | 2 | 1412 | ||
GenBank mRNA | AY250703.1 | - | 2 | 1386 | ||
AY316298.1 | - | 1 | 1260 | |||
BC087723.1 | - | 1 | 1278 | 7320740 |
