siRNA Details
Gene Symbol: Clec4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005899.2 | NP_001005899.2 | 5 | 566 | ||
GenBank mRNA | AY363176.1 | - | 5 | 612 | ||
AY494060.1 | - | 5 | 565 | |||
AY581051.1 | - | 5 | 566 |
