siRNA Details
Gene Symbol: Mib2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005901.2 | NP_001005901.2 | 3 | 1052 | ||
XM_006239588.1 | XP_006239650.1 | 2 | 371 | |||
XM_006239589.2 | XP_006239651.1 | 2 | 333 | |||
XM_008764368.1 | XP_008762590.1 | 2 | 1774 | |||
XM_008764369.1 | XP_008762591.1 | 2 | 1774 | |||
XM_008764370.1 | XP_008762592.1 | 2 | 1774 | |||
XM_008764371.1 | XP_008762593.1 | 2 | 1774 | |||
XM_008764372.1 | XP_008762594.1 | 2 | 1774 |
