siRNA Details
Gene Symbol: Ggnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009972.1 | NP_001009972.1 | 1 | 106 | ||
XM_006256125.2 | XP_006256187.1 | 1 | 152 | |||
GenBank mRNA | AY529104.1 | - | 1 | 106 | ||
BC105838.1 | - | 1 | 114 | 7452960 |
