siRNA Details
Gene Symbol: Plcz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012234.1 | NP_001012234.1 | 3 | 295 | ||
XM_006237597.2 | XP_006237659.1 | 4 | 524 | |||
XM_006237600.2 | XP_006237662.1 | 3 | 400 | |||
GenBank mRNA | AY885259.1 | - | 3 | 295 |
