siRNA Details
Gene Symbol: Scn3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013119.1 | NP_037251.1 | 11 | 1986 | ||
XM_008761914.1 | XP_008760136.1 | 10 | 1789 | |||
XM_008761915.1 | XP_008760137.1 | 11 | 1792 | |||
XM_008761916.1 | XP_008760138.1 | 10 | 1790 | |||
XM_008761917.1 | XP_008760139.1 | 10 | 1789 | |||
XM_008761918.1 | XP_008760140.1 | 10 | 1789 | |||
XM_008761919.1 | XP_008760141.1 | 10 | 1789 | |||
XM_008761920.1 | XP_008760142.1 | 10 | 1789 | |||
GenBank mRNA | Y00766.1 | - | 11 | 1986 |
