siRNA Details
Gene Symbol: Dut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040271.1 | NP_001035361.1 | 5 | 417 | ||
NM_053592.2 | NP_446044.1 | 6 | 537 | |||
GenBank mRNA | AI705003.1 | - | 2 | 406 | ||
CB325379.1 | - | 5 | 431 | |||
FQ220451.1 | - | 572 | ||||
FQ233848.1 | - | 1 | 37 | |||
U64030.2 | - | 6 | 537 |
