siRNA Details
Gene Symbol: Atf7ip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017471.1 | NP_001017471.1 | 6 | 606 | ||
XM_006245728.2 | XP_006245790.1 | 6 | 760 | |||
XM_006245732.2 | XP_006245794.1 | 6 | 726 | |||
XM_006245735.1 | XP_006245797.1 | 4 | 334 | |||
XM_008767495.1 | XP_008765717.1 | 6 | 761 | |||
XM_008767496.1 | XP_008765718.1 | 6 | 759 | |||
GenBank mRNA | BC083663.1 | - | 6 | 606 | 7135436 |
