siRNA Details
Gene Symbol: Nudt16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100782.1 | NP_001094252.1 | 3 | 523 | ||
GenBank mRNA | BC105819.1 | - | 3 | 463 | ||
BC158814.1 | - | 3 | 523 | 7371601 | ||
FQ213246.1 | - | 3 | 513 | |||
FQ214509.1 | - | 3 | 532 | |||
FQ219409.1 | - | 3 | 518 |
