siRNA Details
Gene Symbol: Mrpl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_573060.5 | XP_573060.3 | 3 | 622 | ||
GenBank mRNA | FQ219872.1 | - | 3 | 594 |

More Information
Other Species Targeted

Gene Ontology Categories:
