siRNA Details
Gene Symbol: Fam71b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025031.1 | NP_001020202.1 | 1 | 217 | ||
GenBank mRNA | BC082044.1 | - | 1 | 217 | 7131685 |

More Information
Other Species Targeted

