siRNA Details
Gene Symbol: Sgcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109029.1 | NP_001102499.1 | 2 | 427 | ||
NM_001134826.1 | NP_001128298.1 | 2 | 427 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011891, AK048480, AB024923, AK040974, BC130019, and others.
Gene Ontology Categories:
