siRNA Details
Gene Symbol: Zfp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127558.1 | NP_001121030.1 | 5 | 1054 | ||
XM_008767695.1 | XP_008765917.1 | 6 | 1118 | |||
XM_008767697.1 | XP_008765919.1 | 6 | 1126 | |||
XM_008767698.1 | XP_008765920.1 | 6 | 1079 | |||
XM_008767699.1 | XP_008765921.1 | 6 | 1082 | |||
GenBank mRNA | BC161925.1 | - | 5 | 1054 | 8372872 | |
U78123.1 | - | 1 | 43 |
