siRNA Details
Gene Symbol: Rad51c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129777.1 | NP_001123249.1 | 4 | 693 | ||
XM_006247106.1 | XP_006247168.1 | 4 | 771 | |||
GenBank mRNA | BC166862.1 | - | 4 | 600 |
