siRNA Details
Gene Symbol: Dgke | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039341.1 | NP_001034430.1 | 3 | 883 | ||
XM_006247070.2 | XP_006247132.1 | 3 | 1118 | |||
XM_006247071.2 | XP_006247133.1 | 3 | 1118 | |||
GenBank mRNA | BC110049.1 | - | 3 | 883 | 7316890 |
