siRNA Details
Gene Symbol: Tmem100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017479.1 | NP_001017479.1 | 3 | 483 | ||
XM_008768100.1 | XP_008766322.1 | 2 | 1121 | |||
XM_008768101.1 | XP_008766323.1 | 3 | 980 | |||
XM_008768102.1 | XP_008766324.1 | 3 | 710 | |||
GenBank mRNA | BC092577.1 | - | 3 | 483 | 7124708 | |
FQ212975.1 | - | 3 | 488 |
