siRNA Details
Gene Symbol: Fam117a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109039.1 | NP_001102509.1 | 7 | 993 | ||
XM_006247206.1 | XP_006247268.1 | 7 | 988 | |||
XM_006247207.2 | XP_006247269.1 | 6 | 854 | |||
GenBank mRNA | BC168983.1 | - | 5 | 1058 | 7387439 |
