siRNA Details
Gene Symbol: Arl5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109046.1 | NP_001102516.1 | 4 | 322 | ||
XM_006247396.2 | XP_006247458.1 | 4 | 515 | |||
XM_006247397.2 | XP_006247459.1 | 4 | 509 |
