siRNA Details
Gene Symbol: Dusp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247402.2 | XP_006247464.1 | 4 | 574 | ||
XM_006247403.2 | XP_006247465.1 | 4 | 573 | |||
GenBank mRNA | BC169043.1 | - | 3 | 443 | 9099479 |
