siRNA Details
Gene Symbol: Ern1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191926.1 | NP_001178855.1 | 17, 18 | 2225 | ||
XM_006247633.2 | XP_006247695.1 | 17, 18 | 2476 | |||
XM_006247634.2 | XP_006247696.1 | 17, 18 | 2469 |
