siRNA Details
Gene Symbol: Csnka2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768540.1 | XP_008766762.1 | 1 | 1660 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Csnka2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768540.1 | XP_008766762.1 | 1 | 1660 |