siRNA Details
Gene Symbol: N4bp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035222.2 | NP_001030299.1 | 4 | 429 | ||
XM_008768992.1 | XP_008767214.1 | 4 | 429 | |||
GenBank mRNA | BC168658.1 | - | 4 | 429 | 7314496 | |
FQ231381.1 | - | 4 | 400 | |||
FQ231652.1 | - | 4 | 400 | |||
FQ235206.1 | - | 4 | 400 |
