siRNA Details
Gene Symbol: Spry3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109063.1 | NP_001102533.1 | 2 | 353 | ||
XM_008769065.1 | XP_008767287.1 | 2 | 1727 | |||
XM_008769066.1 | XP_008767288.1 | 3 | 648 | |||
XM_008769067.1 | XP_008767289.1 | 3 | 600 | |||
XM_008769068.1 | XP_008767290.1 | 2 | 316 | |||
XM_008769069.1 | XP_008767291.1 | 2 | 446 | |||
GenBank mRNA | BC168971.1 | - | 2 | 346 | 7316797 |
