siRNA Details
Gene Symbol: Gal3st4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109064.1 | NP_001102534.1 | 4 | 1339 | ||
XM_006249025.2 | XP_006249087.1 | 4 | 1571 | |||
XM_006249026.2 | XP_006249088.1 | 4 | 1357 |
