siRNA Details
Gene Symbol: Prkrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098793.1 | NP_001092263.1 | 5 | 466 | ||
GenBank mRNA | BC158549.1 | - | 5 | 466 | 8361725 | |
BQ194450.1 | - | 2 | 463 | |||
DY314400.1 | - | 5 | 466 |
