siRNA Details
Gene Symbol: Mapkapk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025761.2 | NP_001020932.2 | 2, 3 | 194 | ||
NM_001164043.1 | NP_001157515.1 | 5, 6 | 563 | |||
GenBank mRNA | BC099161.1 | - | 2, 3 | 194 | 7381631 | |
FM032487.1 | - | 5, 6 | 563 |
