siRNA Details
Gene Symbol: Slc8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017488.1 | NP_001017488.1 | 3 | 324 | ||
XM_008769249.1 | XP_008767471.1 | 3 | 367 | |||
GenBank mRNA | BC079350.1 | - | 3 | 324 | 7107820 |
