siRNA Details
Gene Symbol: Fam163a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109072.1 | NP_001102542.1 | 5 | 875 | ||
XM_008769668.1 | XP_008767890.1 | 5 | 1114 | |||
XM_008769669.1 | XP_008767891.1 | 5 | 1111 | |||
XM_008769670.1 | XP_008767892.1 | 5 | 1111 | |||
XM_008769671.1 | XP_008767893.1 | 5 | 955 | |||
XM_008769672.1 | XP_008767894.1 | 3 | 792 |
