siRNA Details
Gene Symbol: RGD1560672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109073.1 | NP_001102543.1 | 2 | 381 | ||
XM_006250221.2 | XP_006250283.1 | 2 | 594 |

More Information
Other Species Targeted

