siRNA Details
Gene Symbol: Nsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109083.1 | NP_001102553.1 | 4, 5 | 532 | ||
XM_006250488.1 | XP_006250550.1 | 4, 5 | 528 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109083.1 | NP_001102553.1 | 4, 5 | 532 | ||
XM_006250488.1 | XP_006250550.1 | 4, 5 | 528 |