siRNA Details
Gene Symbol: Pkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191934.1 | NP_001178863.1 | 6 | 1362 |

More Information
Other Species Targeted

Gene Ontology Categories:

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191934.1 | NP_001178863.1 | 6 | 1362 |