siRNA Details
Gene Symbol: LOC498330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017495.1 | NP_001017495.1 | 4 | 572 | ||
XM_008770050.1 | XP_008768272.1 | 3 | 248 | |||
GenBank mRNA | BC079214.1 | - | 4 | 572 | 7131874 |
Gene Symbol: LOC102546454 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770126.1 | XP_008768348.1 | 200 |
